Merck
  • Disruption of a neural microcircuit in the rod pathway of the mammalian retina by diabetes mellitus.
Disruption of a neural microcircuit in the rod pathway of the mammalian retina by diabetes mellitus.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2015-04-04)
Áurea Castilho, António F Ambrósio, Espen Hartveit, Margaret L Veruki
ABSTRACT
Diabetes leads to dysfunction of the neural retina before and independent of classical microvascular diabetic retinopathy, but previous studies have failed to demonstrate which neurons and circuits are affected at the earliest stages. Here, using patch-clamp recording and two-photon Ca(2+) imaging in rat retinal slices, we investigated diabetes-evoked changes in a microcircuit consisting of rod bipolar cells and their dyad postsynaptic targets, AII and A17 amacrine cells, which play an essential role in processing scotopic visual signals. AII amacrines forward their signals to ON- and OFF-cone bipolar cells and A17 amacrines provide GABAergic feedback inhibition to rod bipolar cells. Whereas Ca(2+)-permeable AMPA receptors mediate input from rod bipolar cells to both AII and A17 amacrines, diabetes changes the synaptic receptors on A17, but not AII amacrine cells. This was expressed as a change in pharmacological properties and single-channel conductance of the synaptic receptors, consistent with an upregulation of the AMPA receptor GluA2 subunit and reduced Ca(2+) permeability. In addition, two-photon imaging revealed reduced agonist-evoked influx of Ca(2+) in dendritic varicosities of A17 amacrine cells from diabetic compared with normal animals. Because Ca(2+)-permeable receptors in A17 amacrine cells mediate synaptic release of GABA, the reduced Ca(2+) permeability of these receptors in diabetic animals leads to reduced release of GABA, followed by disinhibition and increased release of glutamate from rod bipolar cells. This perturbation of neuron and microcircuit dynamics can explain the decreased dynamic range and sensitivity of scotopic vision that has been observed in diabetes.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Potassium D-gluconate, ≥99% (HPLC)
Sigma-Aldrich
Calcium chloride