Merck
  • ¹H-NMR and MS based metabolomics study of the intervention effect of curcumin on hyperlipidemia mice induced by high-fat diet.
¹H-NMR and MS based metabolomics study of the intervention effect of curcumin on hyperlipidemia mice induced by high-fat diet.
PloS one (2015-03-19)
Ze-Yun Li, Li-Li Ding, Jin-Mei Li, Bao-Li Xu, Li Yang, Kai-Shun Bi, Zheng-Tao Wang
ABSTRACT
Curcumin, a principle bioactive component of Curcuma longa L, is well known for its anti-hyperlipidemia effect. However, no holistic metabolic information of curcumin on hyperlipidemia models has been revealed, which may provide us an insight into the underlying mechanism. In the present work, NMR and MS based metabolomics was conducted to investigate the intervention effect of curcumin on hyperlipidemia mice induced by high-fat diet (HFD) feeding for 12 weeks. The HFD induced animals were orally administered with curcumin (40, 80 mg/kg) or lovastatin (30 mg/kg, positive control) once a day during the inducing period. Serum biochemistry assay of TC, TG, LDL-c, and HDL-c was conducted and proved that treatment of curcumin or lovastatin can significantly improve the lipid profiles. Subsequently, metabolomics analysis was carried out for urine samples. Orthogonal Partial Least Squares-Discriminant analysis (OPLS-DA) was employed to investigate the anti-hyperlipidemia effect of curcumin and to detect related potential biomarkers. Totally, 35 biomarkers were identified, including 31 by NMR and nine by MS (five by both). It turned out that curcumin treatment can partially recover the metabolism disorders induced by HFD, with the following metabolic pathways involved: TCA cycle, glycolysis and gluconeogenesis, synthesis of ketone bodies and cholesterol, ketogenesis of branched chain amino acid, choline metabolism, and fatty acid metabolism. Besides, NMR and MS based metabolomics proved to be powerful tools in investigating pharmacodynamics effect of natural products and underlying mechanisms.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Allantoin, ≥98.0% (N)
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Supelco
Acetone, analytical standard
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
DL-3-Hydroxy-3-methylglutaryl coenzyme A sodium salt hydrate, ≥90% (HPLC)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
PalGly, ≥98% (HPLC), solid
Supelco
Allantoin, analytical standard
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Supelco
Allantoin, Pharmaceutical Secondary Standard; Certified Reference Material
Valine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard