Merck
  • The chemical profile of pyrrolizidine alkaloids from selected greek endemic boraginaceae plants determined by gas chromatography/mass spectrometry.
The chemical profile of pyrrolizidine alkaloids from selected greek endemic boraginaceae plants determined by gas chromatography/mass spectrometry.
Journal of AOAC International (2015-04-24)
Harilaos Damianakos, Malgorzata Jeziorek, Agnieszka Pietrosiuk, Ioanna Chinou
ABSTRACT
Four Greek endemic Boraginaceae plants, Onosma erecta Sibth. & Sm., Onosma kaheirei Teppner, Onosma leptantha Heldr., and Cynoglossum columnae L. (aerial parts), were screened for their content of pyrrolizidine alkaloids (PAs). TLC with the Mattocks-Molyneux visualization reagent was used as a preliminary qualitative test for PA or PA N-oxide detection. The extracts of the species found to contain PAs and their N-oxides were further analyzed by GC/MS, so as to identify their structures by means of the mass spectra and retention index values of known PAs already published in the literature. Twenty-three PAs were identified. For additional peaks, recognized as possible PAs by their MS pattern, no exact structures were tentatively suggested, as a result of lack of matching literature data. Furthermore, a quantitative PA profile of the species was obtained.
Product Description
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Acetic anhydride, 99.5%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Supelco
Acetic anhydride, derivatization grade (GC derivatization), LiChropur, ≥99.0%
Supelco
Toluene, analytical standard
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Acetic anhydride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Vanillin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
3,4,5,6-Tetrachloro-1,2-benzoquinone, 97%
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
Vanillin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%