Merck
  • Quantification of serum IgG subclasses by use of subclass-specific tryptic peptides and liquid chromatography--tandem mass spectrometry.
Quantification of serum IgG subclasses by use of subclass-specific tryptic peptides and liquid chromatography--tandem mass spectrometry.
Clinical chemistry (2014-05-07)
Paula M Ladwig, David R Barnidge, Melissa R Snyder, Jerry A Katzmann, David L Murray
ABSTRACT
Measurement of IgG subclasses is a useful tool for investigation of humoral immune deficiency in the presence of total IgG within reference intervals and IgG4-related disease. Nephelometry has been the method of choice for quantification. We describe an LC-MS/MS method that can multiplex all 4 subclasses along with total IgG by use of either IgG subclass-specific peptide stable isotope-labeled internal standards or a surrogate digest standard for quantification and does not rely on antigen/antibody reactions. We combined serum with labeled internal peptide standards and intact purified horse IgG. Samples were denatured, reduced, alkylated, and digested. We analyzed the digested serum by LC-MS/MS for IgG subclasses 1-4 and total IgG. We assayed 112 patient sera by LC-MS/MS and immunonephelometry. The mean of the slopes and R(2) values for IgG1, IgG2, IgG3, IgG4, and IgG were 1.18 and 0.93, respectively. Interassay imprecision for the LC-MS/MS method was <15% for total IgG and subclasses and was slightly improved by use of a calibrator peptide from an exogenous horse IgG. Summed total IgG correlated with the measured total IgG within 10%. Reference intervals and analytical measuring range were all similar to our previous validation data for the immunonephelometry assays. Total IgG and IgG subclasses 1, 2, 3, and 4 can be quantified by LC-MS/MS with performance comparable to nephelometry.
Product Description
Supelco
2,2,2-Trifluoroethanol, analytical standard, suitable for NMR (spectroscopy)
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2,2,2-Trifluoroethanol, ReagentPlus®, ≥99%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
SAFC
Iodoacetamide