Merck
  • Peripheral neurobiologic mechanisms of antiallodynic effect of warm water immersion therapy on persistent inflammatory pain.
Peripheral neurobiologic mechanisms of antiallodynic effect of warm water immersion therapy on persistent inflammatory pain.
Journal of neuroscience research (2014-08-01)
Daniel F Martins, Rômulo N Brito, Juliana Stramosk, Ana P Batisti, Fernanda Madeira, Bruna L Turnes, Leidiane Mazzardo-Martins, Adair R S Santos, Anna P Piovezan
ABSTRACT
Water immersion is widely used in physiotherapy and might relieve pain, probably by activating several distinct somatosensory modalities, including tactile, pressure, and thermal sensations. However, the endogenous mechanisms behind this effect remain poorly understood. This study examined whether warm water immersion therapy (WWIT) produces an antiallodynic effect in a model of localized inflammation and whether peripheral opioid, cannabinoid, and adenosine receptors are involved in this effect. Mice were injected with complete Freund's adjuvant (CFA; intraplantar; i.pl.). The withdrawal frequency to mechanical stimuli (von Frey test) was used to determine 1) the effect of WWIT against CFA-induced allodynia and 2) the effect of i.pl. preadministration of naloxone (a nonselective opioid receptor antagonist; 5 µg/paw), caffeine (a nonselective adenosine receptor antagonist; 150 nmol/paw), 1,3-dipropyl-8-cyclopentylxanthine (DPCPX; a selective adenosine A1 receptor antagonist; 10 nmol/paw), and AM630 (a selective cannabinoid receptor type 2 antagonist; 4 µg/paw) on the antiallodynic effect of WWIT against CFA-induced allodynia. Moreover, the influence of WWIT on paw inflammatory edema was measured with a digital micrometer. WWIT produced a significant time-dependent reduction of paw inflammatory allodynia but did not influence paw edema induced by CFA. Naloxone, caffeine, DPCPX, and AM630 injected in the right, but not in the left, hind paw significantly reversed the antiallodynic effect of WWIT. This is the first study to demonstrate the involvement of peripheral receptors in the antiallodynic effect of WWIT in a murine model of persistent inflammatory pain.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
8-Cyclopentyl-1,3-dipropylxanthine, solid
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
AM630, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Adenosine, European Pharmacopoeia (EP) Reference Standard
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous