Merck
  • Loss of calpastatin leads to activation of calpain in human lens epithelial cells.
Loss of calpastatin leads to activation of calpain in human lens epithelial cells.
Investigative ophthalmology & visual science (2014-07-24)
Takeshi Nakajima, Thomas R Shearer, Mitsuyoshi Azuma
ABSTRACT
Activation of calpains (calpain 2 and Lp82) in rodent lenses readily causes proteolysis and cataract formation. In contrast, primate lenses are quite resistant to activation of calpains. The hypothesis is that high levels of human endogenous calpain inhibitor, calpastatin (CS), prevent calpain activation in human lenses. The purpose of the present study was to directly test if CS is a major inhibitory factor in a human lens epithelial cell line, HLE B-3. Small interfering RNAs (siRNAs) were used to knock down expression of CS in HLE B-3. The cells then were cultured with the calcium ionophore, ionomycin, with or without a calpain inhibitor SNJ-1945. Transcripts for calpain 2 and CS were measured by quantitative PCR (qPCR). Calpain 2 activity was detected by immunoblotting for the calpain-specific, α-spectrin breakdown product and for activation-associated, fragments of calpain 2. Expression of CS in HLE B-3 was remarkably higher than in α-TN4 (mouse comparator cell line). Proteolysis of α-spectrin was observed in the soluble proteins from α-TN4 incubated with Ca(2+), but not in the human HLE B-3. When CS-reduced HLE B-3 cells (transfected with CS siRNA) were cultured with ionomycin, calpain 2 was activated, specific proteolysis of α-spectrin occurred, and cell death ensued; SNJ-1945 inhibited these changes. Our data demonstrated that the high levels of endogenous CS do, indeed, inhibit calpain activity in normal human lens epithelial cells. We speculate that age-related oxidation might cause loss of CS activity in human lens epithelial cells, allowing activation of long-dormant calpain 2, proteolysis of critical cytoskeletal proteins, and cataract formation.
Sigma-Aldrich
MISSION® esiRNA, targeting human CS
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer