Merck
Axl-targeted cancer imaging with humanized antibody h173.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging (2014-01-16)
Dan Li, Shuanglong Liu, Ren Liu, Ryan Park, Haiyang Yu, Valery Krasnoperov, Parkash S Gill, Zibo Li, Hong Shan, Peter S Conti
ABSTRACT
The tyrosine kinase receptor Axl is overexpressed in various types of cancer and correlated with cancer malignancy. Selective Axl blockade reduces tumor growth and metastasis. The purpose of this study was to examine whether the humanized anti-Axl antibody humanized 173 (h173) labeled with near-infrared fluorescence (NIRF) dye Cy5.5 could be applied as a molecular imaging probe for NIRF imaging of Axl expression in tumor models. NIRF dye Cy5.5 was conjugated to h173 or human normal immunoglobulin G (hIgG) control through amino groups. The resulting probes were evaluated in both A549 (Axl positive) and NCI-H249 (Axl negative) lung cancer xenografts through in vivo NIRF imaging. Ex vivo imaging and probe distribution assay were also carried out to confirm the in vivo imaging results. After conjugation, binding activity of h173-Cy5.5 was determined to be 97.75 % ± 2.09 % of the unmodified h173. In vitro fluorescence-activated cell sorting (FACS) and fluorescence microscopy analysis validated the specific binding of h173 toward Axl-positive A549 cells. h173-Cy5.5 was then applied to image Axl expression in vivo. In A549 (Axl positive) cancer xenografts, the tumor uptake of h173-Cy5.5 was significantly higher than that of the hIgG-Cy5.5 control (P < 0.05) at late time points (1, 2, 3, 4, and 7 days). On the contrary, in NCI-H249 (Axl negative) cancer xenografts, the tumor uptake of both hIgG-Cy5.5 and h173-Cy5.5 was low and showed no significant difference (P > 0.05) at all time points examined. Ex vivo imaging and immunofluorescence staining analysis further validated the in vivo imaging results. Collectively, all in vitro, in vivo, and ex vivo data suggested that h173-Cy5.5 could serve as a valid probe for Axl-targeted cancer imaging, which could therefore aid in tumor diagnosis, prognosis, and treatment monitoring.
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
5(6)-Carboxyfluorescein, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein, Dye content 90 %