Merck
  • A ligand field series for the 4f-block from experimental and DFT computed Ce(IV/III) electrochemical potentials.
A ligand field series for the 4f-block from experimental and DFT computed Ce(IV/III) electrochemical potentials.
Inorganic chemistry (2015-02-25)
Justin A Bogart, Andrew J Lewis, Michael A Boreen, Heui Beom Lee, Scott A Medling, Patrick J Carroll, Corwin H Booth, Eric J Schelter
ABSTRACT
Understanding of the sensitivity of the reduction potential of cerium(IV) cations to ligand field strength has yet to benefit from systematic variation of the ligand environment. Detailed analyses for a series of seven cerium(IV) tetrakis(pyridyl-nitroxide) compounds and their cerium(III) analogues in varying ligand field strengths are presented. Electrochemical, spectroscopic, and computational results reveal a close correlation of electronic properties with ligand substituents. Together with electrochemical data for reported eight-coordinate compounds, DFT calculations reveal a broad range of the cerium(IV/III) redox potentials correlated to ligand field strengths, establishing a semiempirical, predictive model for the modulation of cerium redox thermodynamics and ligand field strengths. Applications over a variety of scientific disciplines make use of the fundamental redox thermodynamics of cerium. Such applications will benefit from a combined experimental and theoretical approach for assessing redox cycling of cerium compounds.
Sigma-Aldrich
Pentane, ≥99% (GC)
Sigma-Aldrich
Pentane, spectrophotometric grade, ≥99%
Sigma-Aldrich
Pentane, reagent grade, 98%
Sigma-Aldrich
Pentane, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Pentane, suitable for HPLC, ≥99.0%
Sigma-Aldrich
2-Bromopyridine, 99%
Sigma-Aldrich
Pentane, anhydrous, ≥99%
Supelco
Pentane, analytical standard
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
1,2-Dimethoxyethane, anhydrous, 99.5%, inhibitor-free
Supelco
1,2-Dimethoxyethane, analytical standard
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
1,2-Dimethoxyethane, ReagentPlus®, ≥99%, inhibitor-free
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, ≥99.0%, contains 200-400 ppm BHT as inhibitor, ReagentPlus®
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%