Merck
  • The effect of etoricoxib on kidney ischemia-reperfusion injury in rats: a biochemical and immunohistochemical assessment.
The effect of etoricoxib on kidney ischemia-reperfusion injury in rats: a biochemical and immunohistochemical assessment.
International immunopharmacology (2014-07-30)
Bahadir Suleyman, Abdulmecit Albayrak, Nezahat Kurt, Elif Demirci, Cemal Gundogdu, Mehmet Aksoy
ABSTRACT
The purpose of this study was to investigate the effect of etoricoxib on oxidative injury induced with ischemia-reperfusion (I/R) in rat kidney tissue in terms of biochemistry and immunohistochemistry. Male Albino Wistar rats were divided into renal I/R (RIR), 50 mg/kg etoricoxib+RIR (ETO-50), 100 mg/kg etoricoxib+RIR (ETO-100) and sham operation (SG) groups. Animals in the ETO-50 and ETO-100 groups were given etoricoxib by the oral route at dosages of 50 and 100 mg/kg, respectively. The RIR and SG groups were given distilled water as solvent. One hour after drug administration, 1h of ischemia and 3h of reperfusion were applied to the left kidneys of all rats (apart from SG) under 25 mg/kg thiopental sodium anesthesia. At the end of that time, kidneys were extracted and biochemical and immunohistochemical analyses were performed. Etoricoxib reduced, in a dose-dependent manner, levels of MDA, MPO and COX-2 that normally rise with I/R in rat kidney tissues. Etorixicob did not alter COX-1 activity at 50 and 100 mg/kg doses, but significantly prevented loss of tGSH in tissues with I/R. In addition, Bcl-2' gene expression inhibited with I/R was prevented in renal tubular and glomerular cells. Furthermore, etoricoxib significantly decreased the caspase-3 gene expression which increased with I/R. Etoricoxib significantly prevented I/R injury in a dose-dependent manner. The results of this study show that etoricoxib treatment could decrease kidney injury during IR.