Merck
Unexpected photoluminescence of fluorinated naphthalene diimides.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-02-03)
Subashani Maniam, Rosalind P Cox, Steven J Langford, Toby D M Bell
ABSTRACT
Two new amino core-substituted naphthalene diimides (cNDIs) bearing fluorinated side chains have been synthesised. Steady-state and time-resolved fluorescence spectroscopy reveals unprecedented optical properties for the cNDIs with high quantum yields of ∼0.8 and fluorescence lifetimes of ∼13 ns in a range of solvents. These properties are apparent at the level of single molecules, where the compounds also show exceptional photostability under pulsed-laser excitation. Photon emission is remarkably consistent with very few long timescale (millisecond or longer) interruptions with molecules regularly undergoing >10(7) cycles of excitation and emission. Intermittencies owing to triplet-state formation occur on a sub-millisecond timescale with a low yield of 1-2%, indicating that the presence of the fluorine atoms does not lead to a significant triplet yield through the heavy-atom effect. These properties make the compounds excellent candidates for single-molecule labelling applications.
Product Description
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Hexylamine, 99%
Sigma-Aldrich
1,4,5,8-Naphthalenetetracarboxylic dianhydride
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Supelco
Toluene, analytical standard
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Benzonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)