Merck
  • Genotoxicity of chlorpyrifos in freshwater fish Labeo rohita using Alkaline Single-cell Gel Electrophoresis (Comet) assay.
Genotoxicity of chlorpyrifos in freshwater fish Labeo rohita using Alkaline Single-cell Gel Electrophoresis (Comet) assay.
Drug and chemical toxicology (2014-02-15)
Muhammad Ismail, Qaiser Mahmood Khan, Rahat Ali, Tayyaba Ali, Ameena Mobeen
ABSTRACT
Chlorpyrifos is a widely used insecticide of organophosphate group, which causes severe toxicological effects in non target aquatic organisms especially in fish. In the present study the genotoxic effects of sublethal concentrations of chlorpyrifos were observed in the erythrocytes and gill cells of Labeo rohita (commonly known as rohu) using the Alkaline Single-Cell Gel Electrophoresis (Comet) assay. Effects of chlorpyrifos on the behavior of the fish were also investigated. The 96 h LC50 value of chlorpyrifos, estimated by Trimmed Spearman-Karber (TSK) in static bioassay, was found to be 442.8 µg/L. On the basis of LC50 value, the fish were exposed to three sublethal concentrations of chlorpyrifos (SL-I ∼221.4 µg/L, SL- II ∼110.7 µg/L and SL-III ∼73.8 µg/L) for 96 h. Blood and gill samples were collected at every 24 h and were subjected to the Comet assay. The observed DNA damage was concentration dependent and time dependent and those levels of DNA damage in between the tested concentrations and times were significantly different (p < 0.01). It was also found that the gill cells are more sensitive to chlorpyrifos, though; it revealed more DNA damage as compared to the erythrocytes of fish. Fish exposed to different concentrations of chlorpyrifos showed different neurotoxic behavioral responses. It was concluded that chlorpyrifos is a genotoxic and neurotoxic insecticide causing DNA damage and neurotoxic effects in Labeo rohita.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Supelco
Methyl methanesulfonate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Methyl methanesulfonate, 99%
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O