Merck
  • Enhanced oral delivery of curcumin from N-trimethyl chitosan surface-modified solid lipid nanoparticles: pharmacokinetic and brain distribution evaluations.
Enhanced oral delivery of curcumin from N-trimethyl chitosan surface-modified solid lipid nanoparticles: pharmacokinetic and brain distribution evaluations.
Pharmaceutical research (2014-08-02)
Prakash Ramalingam, Young Tag Ko
ABSTRACT
Solid lipid nanoparticles (SLNs) have been proposed as a colloidal carrier system that could enhance the oral bioavailability of curcumin. However, a burst release of the loaded drug, which occurs in acidic environments, has been a main obstacle to the oral delivery of curcumin by using SLNs as a carrier system. We hypothesized that a quarternized chitosan derivative could be used for acid-resistant coating to stabilize the SLNs and circumvent the burst release. N-trimethyl chitosan (TMC) was synthesized and determined by (1)H-NMR and FT-IR. To investigate the details of chitosan and TMC surface modification on SLCNs composed of palmitic acid, cholesterol, TPGS and curcumin, a number of factors such as optimized SLNs composition, solid state characterization, stability, cell viability, in vitro release in GI conditions, curcumin oral bioavailability and brain distribution studies, were evaluated. The TMC-SLCNs exhibited prolonged stability in room and refrigerated conditions, controlled drug release in simulated intestinal fluid, significantly higher oral bioavailability, and brain distribution of curcumin than free curcumin, chitosan and non-coated SLCNs. These finding suggests that the TMC-SLCNs is a promising nanocarrier system for oral delivery and brain distribution of curcumin.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Supelco
1-Methyl-2-pyrrolidinone, analytical standard
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder