Merck
  • Cyclic sulfoxides-garlicnins K1, K2, and H1-extracted from Allium sativum.
Cyclic sulfoxides-garlicnins K1, K2, and H1-extracted from Allium sativum.
Chemical & pharmaceutical bulletin (2015-03-10)
Toshihiro Nohara, Yukio Fujiwara, Yusuke Komota, Yoshihiko Kondo, Taiki Saku, Koki Yamaguchi, Yoshihiro Komohara, Motohiro Takeya
ABSTRACT
Newly identified cyclic sulfoxides-garlicnins K1 (1), K2 (2), and H1 (3)-were isolated from the acetone extracts of the bulbs of garlic, Allium sativum. Garlicnin H1 (3) demonstrated potential to suppress tumor cell proliferation by regulating macrophage activation. The structures of garlicnins K1 and K2, 3,4-dimethyl-5-allyl-tetrahydrothiophen-2-one-S-oxides, and the structure of garlicnin H1, 3-carboxy-3-hydroxy-4-methyl-5-allylsulfoxide-tetrahydrothiophen-2-(ethane-1,2-diol)-S-oxide were characterized by spectroscopic analysis.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)