Merck
  • Glycine improves biochemical and biomechanical properties following inflammation of the achilles tendon.
Glycine improves biochemical and biomechanical properties following inflammation of the achilles tendon.
Anatomical record (Hoboken, N.J. : 2007) (2014-08-27)
Cristiano Pedrozo Vieira, Letícia Prado De Oliveira, Flávia Da Ré Guerra, Marcos Dos Santos De Almeida, Maria Cristina Cintra Gomes Marcondes, Edson Rosa Pimentel
ABSTRACT
Tendinopathy of the Achilles tendon is a clinical problem that motivates the scientific community to search for treatments that assist in restoring its functional properties. Glycine has broad biological effects, acting as a modulator of the inflammatory cascade, and is the predominant amino acid in collagen. A 5% glycine diet provided beneficial effects against toxicity and inflammation since glycine may restructure the collagen molecules faster due to its broad anti-inflammatory effects. The purpose was analyze the effects of a 5% glycine diet in rats as a treatment for the inflammatory process. The experimental groups were as follows: C (control group), G1 and G3 (inflammatory group), and G2 and G4 (glycine+inflammatory group). G1 and G2 were euthanized 8 days following injury, and G3 and G4 were euthanized 22 days following injury. The concentrations of hydroxyproline, non-collagenous proteins, and glycosaminoglycans, as well as the activity of MMP-2 and -9 were analyzed. Biomechanical and morphological tests were employed. Higher concentrations of hydroxyproline and glycosaminoglycans were found in G4 and an increased activity of MMP-2 was found in G2. Higher birefringence was noted in group G2. The biomechanical results indicated that the tendon was more resistant to loading to rupture upon treatment with a glycine diet in group G4. Glycine induced the synthesis of important components of the tendon. A rapid remodeling was noted when compared with the inflamed-only groups. These data suggest that glycine may be a beneficial supplement for individuals with inflammation of the Achilles tendon.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
SAFC
Glycine
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Glycine, European Pharmacopoeia (EP) Reference Standard
USP
Glycine, United States Pharmacopeia (USP) Reference Standard