Merck
  • New phenolic compounds from Coreopsis tinctoria Nutt. and their antioxidant and angiotensin i-converting enzyme inhibitory activities.
New phenolic compounds from Coreopsis tinctoria Nutt. and their antioxidant and angiotensin i-converting enzyme inhibitory activities.
Journal of agricultural and food chemistry (2014-12-18)
Wei Wang, Wei Chen, Yingshi Yang, Tianxing Liu, Haiyan Yang, Zhihong Xin
ABSTRACT
Three new phenolic compounds, coretinphenol (1), coretincone (2), and coretinphencone (3), were isolated from the buds of Coreopsis tinctoria Nutt., together with nine known compounds, including butein (4), okanin (5), isoliquiritigenin (6), maritimetin (7), taxifolin (8), isookanin (9), marein (10), sachalinoside B (11), and 2-phenylethyl-β-d-glucoside (12). The chemical structures of these compounds were elucidated by extensive spectroscopic analysis and on the basis of their chemical reactivity. This work represents the first recorded example of the isolation of compounds 1–3, 6, 7, 9, 11, and 12 from C. tinctoria. Compounds 5–9 showed strong diphenyl(2,4,6-trinitrophenyl)iminoazanium (DPPH) radical-scavenging activity, with IC50 values of 3.35 ± 0.45, 9.6 ± 2.32, 4.12 ± 0.21, 6.2 ± 0.43, and 7.9 ± 0.53 μM, respectively. Compounds 2 and 8 exhibited angiotensin I-converting enzyme inhibitory activity, with IC50 values of 228 ± 4.47 and 145.67 ± 3.45 μM, respectively. The activities of phenolic compounds isolated from C. tinctoria support the medicinal use of this plant in the prevention of cardiovascular diseases.
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Hippuric acid, 98%
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Captopril for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Supelco
Hexane, analytical standard
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Captopril, meets USP testing specifications