Merck
  • Ligands for FKBP12 increase Ca2+ influx and protein synthesis to improve skeletal muscle function.
Ligands for FKBP12 increase Ca2+ influx and protein synthesis to improve skeletal muscle function.
The Journal of biological chemistry (2014-07-24)
Chang Seok Lee, Dimitra K Georgiou, Adan Dagnino-Acosta, Jianjun Xu, Iskander I Ismailov, Mark Knoblauch, Tanner O Monroe, RuiRui Ji, Amy D Hanna, Aditya D Joshi, Cheng Long, Joshua Oakes, Ted Tran, Benjamin T Corona, Sabina Lorca, Christopher P Ingalls, Vihang A Narkar, Johanna T Lanner, J Henri Bayle, William J Durham, Susan L Hamilton
ABSTRACT
Rapamycin at high doses (2-10 mg/kg body weight) inhibits mammalian target of rapamycin complex 1 (mTORC1) and protein synthesis in mice. In contrast, low doses of rapamycin (10 μg/kg) increase mTORC1 activity and protein synthesis in skeletal muscle. Similar changes are found with SLF (synthetic ligand for FKBP12, which does not inhibit mTORC1) and in mice with a skeletal muscle-specific FKBP12 deficiency. These interventions also increase Ca(2+) influx to enhance refilling of sarcoplasmic reticulum Ca(2+) stores, slow muscle fatigue, and increase running endurance without negatively impacting cardiac function. FKBP12 deficiency or longer treatments with low dose rapamycin or SLF increase the percentage of type I fibers, further adding to fatigue resistance. We demonstrate that FKBP12 and its ligands impact multiple aspects of muscle function.
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%
Supelco
2-Methylbutane, analytical standard
Sigma-Aldrich
2-Methylbutane, suitable for HPLC, ≥99.5%
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, puriss. p.a., ≥99.5% (GC)
Supelco
Rapamycin, VETRANAL®, analytical standard