Merck
  • Production, HPLC analysis, and in situ apoptotic activities of swainsonine toward lepidopteran, Sf-21 cell line.
Production, HPLC analysis, and in situ apoptotic activities of swainsonine toward lepidopteran, Sf-21 cell line.
Biotechnology progress (2014-07-22)
Digar Singh, Gurvinder Kaur
ABSTRACT
Swainsonine, a secondary metabolite from Metarhizium anisopliae has been extensively studied in the complementary areas of therapeutics and toxicology. This work aims to develop a simple UV-HPLC method of analyses for swainsonine in Metarhizium fermentation broth and to explore its in situ entomotoxic activities. The partially purified broth was quantitatively analyzed using middle UV (205 nm)-reverse phase HPLC method with different mobile phases and gradient programmes. Swainsonine was eluted as single peak at (te ) 6.0-6.9 min with average concentration of 4.04 ± 0.52 μg/mL using optimal mobile phase (0.1% trifluoroacetic acid in water and acetonitrile). The mass spectrometry analysis further indicated the characteristic MS1 species for swainsonine, [M+H](+) 174.30 in corresponding HPLC peaks. The antiproliferative effects of swainsonine on lepidopteran, Sf-21 cells were determined through 3-(4, 5-dimethylthia-zol-2-yl)-2, 5-diphenyl tetrazolium bromide (IC50 standard  = 3.90 μM and IC50 purified  = 5.27 μM) and trypan blue dye exclusion (IC50 standard  = 6.91 μM and IC50 purified  = 8.67 μM) assays. The fluorescence activated cell sorting evaluation of Sf-21 cells showed nearly 35% and 42% of population in various apoptotic stages at 36 h, when treated with standard and purified swainsonine, respectively. The morphodimensional field emission scanning electron and atomic force microscopic analyses further confirmed the characteristic apoptotic features like membrane blebbings, ruptures and volume shrinkage in the lepidopteran cells after 24-36 h of post-treatment incubation. The study describes the potential entomotoxic activities of swainsonine and its role in the virulence of Metarhizium spp.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%