Merck
  • Stability-indicating determination of trimetazidine dihydrochloride inthe presence of two of its related substances using a direct GC/MS method.
Stability-indicating determination of trimetazidine dihydrochloride inthe presence of two of its related substances using a direct GC/MS method.
Journal of AOAC International (2015-01-30)
Tarek S Belal, Tamer Awad, C Randall Clark
ABSTRACT
A novel, simple, direct, and selective stability- indicating GC/MS procedure was developed for the determination of the anti-ischemic drug trimetazidine dihydrochloride (TMZ) in the presence of two of its related substances (potential impurities), namely, 2,3,4-trimethoxybenzyl alcohol (T1) and 2,3,4-trimethoxybenzaldehyde (T2). The method involved resolution of the undeilvatized compounds using a 100% dimethylpolysiloxane (Rtx-1) capillary column, and MS detection was carried out in the electron-impact mode. The peaks of the three compounds eluted at retention times 11.69, 11.92, and 15.47 min for T1, T2, and TMZ, respectively. Quantification of the parent drug TMZ was based on measuring its peak area. The reliability and analytical performance of the proposed method, including linearity, range, precision, accuracy, selectivity, detection, and quantification limits, were statistically validated. The calibration curve of TMZ was linear over the range 100-600 μg/mL. The proposed method was successfully applied to the assay of TMZ in several commercially available pharmaceutical formulations with recoveries not lessthan 96.2%.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%