Merck
  • Polymer hydrogel functionalized with biodegradable nanoparticles as composite system for controlled drug delivery.
Polymer hydrogel functionalized with biodegradable nanoparticles as composite system for controlled drug delivery.
Nanotechnology (2014-12-10)
Filippo Rossi, Raffaele Ferrari, Franca Castiglione, Andrea Mele, Giuseppe Perale, Davide Moscatelli
ABSTRACT
The possibility to direct pharmacological treatments targeting specific cell lines using polymer nanoparticles is one of the main novelties and perspectives in nanomedicine. However, sometimes, the ability to maintain NPs localized at the site of the injection that work as a drug reservoir can represent a good and complementary option. In this direction we built a composite material made of polymeric hydrogel functionalized with polymer NPs. ϵ-caprolactone and polyethylene glycol have been copolymerized in a two-step synthesis of PEGylated NPs, while hydrogel was synthesized through polycondensation between NPs, agarose and branched polyacrylic acid. NP functionalization was verified with Fourier transform infrared spectroscopy (FTIR), high resolution magic angle spinning-nuclear magnetic resonance (HRMAS-NMR) spectroscopy and release kinetics from a hydrogel matrix and compared with NPs only physically entrapped into a hydrogel matrix. The characteristics of the resulting composite hydrogel-NPs system were studied both in terms of rheological properties and in its ability to sustain the release of To-Pro3, used as a drug mimetic compound to represent a promising drug delivery device.
Product Description
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, ≥99.0%, contains 200-400 ppm BHT as inhibitor, ReagentPlus®
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%