Merck
  • Dietary L-arginine modulates immunosuppression in broilers inoculated with an intermediate strain of infectious bursa disease virus.
Dietary L-arginine modulates immunosuppression in broilers inoculated with an intermediate strain of infectious bursa disease virus.
Journal of the science of food and agriculture (2014-04-15)
Jian-Zhuang Tan, Yu-Ming Guo, Todd J Applegate, En-Cun Du, Xu Zhao
ABSTRACT
The effects of dietary l-arginine (Arg) on immunosuppression following infectious bursal disease virus (IBDV) inoculation in broiler chickens were evaluated. The design of this study was a 5 × 2 factorial arrangement (n = 5) with five Arg concentrations (starter: 9.9, 13.9, 17.6, 21.3 and 25.3 g kg(-1) ; grower-finisher: 9.5, 13.5, 17.1, 20.1 and 23.6 g kg(-1) ) with or without IBDV inoculation (IBDV or saline inoculation at 14 days). Chickens were sampled at 2, 4 and 6 days post-inoculation (DPI) and 42 days of age. The IBDV inoculation decreased (P = 0.05) CD3(+) , CD4(+) , and CD8(+) T cell counts at 2 days post-inoculation (DPI) and monocyte counts at 6 DPI; and reduced (P < 0.05) bursal interleukin-1β (IL-1β) mRNA expression at 2 DPI and serum IL-6 concentration at 4 DPI. Increasing Arg concentration increased (P < 0.05) CD4(+) and CD8(+) T cell counts at 2 DPI, linearly increased (P = 0.05) CD3(+) T cell counts in IBDV-inoculated groups and monocyte counts in control groups at 4 DPI; increased (P < 0.05) serum IL-6 concentration in IBDV-inoculated groups at 2 DPI; and increased (P < 0.05) serum anti-IBDV antibody titres at 42 days of age. Varying concentrations of Arg supplementation attenuated IBDV inoculation induced immunosuppression via modulating circulating T cell sub-populations.
Product Description
Supelco
Menadione, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Menadione, European Pharmacopoeia (EP) Reference Standard
DL-Methionine, European Pharmacopoeia (EP) Reference Standard
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
Cyanocobalamin, British Pharmacopoeia (BP) Assay Standard
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Choline chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Methionine, European Pharmacopoeia (EP) Reference Standard
Supelco
Cyanocobalamin (B12), analytical standard
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Riboflavin (B2), analytical standard
Supelco
Menadione (K3), analytical standard
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Folic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
DL-Methionine, ≥99.0% (NT)
SAFC
L-Arginine
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
Choline chloride, ≥99%