Merck
  • Transglutaminase 2 is involved in homocysteine-induced activation of human THP-1 monocytes.
Transglutaminase 2 is involved in homocysteine-induced activation of human THP-1 monocytes.
Free radical research (2014-12-31)
M Currò, C Gangemi, A Gugliandolo, R Risitano, N Ferlazzo, R Ientile, D Caccamo
ABSTRACT
Aberrant transglutaminase 2 (TG2) expression and protein cross-linking activity have been associated with several chronic neurodegenerative disorders in which inflammatory processes triggered by activated microglia and monocytes play a key role, such as Alzheimer's disease, Parkinson's disease, amyotrophic lateral sclerosis, and multiple sclerosis. Interestingly, mild-to-moderate hyperhomocysteinemia (HHcy), corresponding to increased plasma homocysteine (Hcy) concentrations in the range 16-60 μM, have recently been associated with the above-cited diseases. Using THP-1 monocytes, here we investigated the role of TG2 in cell response to mildly elevated Hcy concentrations. A five-day incubation with Hcy (∼25 μM) increased reactive oxygen species, peroxide lipids, as well as 8-hydroxyguanosine levels by twofold, and decreased the endogenous cell antioxidant defenses, that is reduced glutathione, by 50% in Hcy-exposed cultures compared with controls (p < 0.01). Hcy-induced oxidative stress was associated with increases in TG2 expression and activity, as well as nuclear factor kappa B activation. Notably, the latter was reduced in the presence of the TG-specific inhibitor R283. Hcy exposure also significantly increased the mRNA levels of tumor necrosis factor alpha, interleukin (IL)-6, and IL-1β, as well as the level of Hcy-inducible endoplasmic reticulum (ER) stress protein, a marker of ER stress, in Hcy-exposed cultures compared with controls. Notably, these effects were dramatically reduced by R283. These preliminary findings indicate that TG2 plays a key role in Hcy-induced activation of THP-1 monocytes, involving oxidative as well as ER stress and inflammation. This underlines the potential of TG2 inhibition in the therapeutic management of inflammatory processes contributing to neurodegenerative disorders associated with mild HHcy.
Sigma-Aldrich
Amylamine, ≥99%, FG
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Amylamine, 99%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Amylamines, mixture of isomers, ≥98%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)