Merck
  • Etoricoxib in the treatment of Korean patients with osteoarthritis in a double-blind, randomized controlled trial.
Etoricoxib in the treatment of Korean patients with osteoarthritis in a double-blind, randomized controlled trial.
Current medical research and opinion (2014-08-19)
Myung Chul Yoo, Wan Hee Yoo, Seung Baek Kang, Yong-Wook Park, Sung Soo Kim, Kyoung Ho Moon, Yeong Wook Song, Byung Woo Min, Yoon Je Cho, Seong-Hwan Moon, Seong-Il Bin, Han-Joo Baek, Seung Cheol Shim, Sung Won Lee, Dae Hyun Yoo, Anish Mehta, Aleksandar Skuban, Diane M Cukrow, Kristel Vandormael, Li Yan
ABSTRACT
We evaluated the COX-2 inhibitors, etoricoxib and celecoxib, in Korean patients with osteoarthritis (OA). This study included patients (≥ 40 years of age) with a clinical and radiographic diagnosis of knee OA. Patients were randomized to etoricoxib 30 mg (qd) or celecoxib 200 mg (qd) in a 12 week randomized, controlled, double-blind study. Prior NSAID users were to demonstrate a worsening of symptoms upon withdrawal of medication. Efficacy endpoints included the time-weighted average change from baseline in the WOMAC VA 3.0 Pain Subscale (100 mm Visual Analog Scale [VAS]; primary endpoint), the WOMAC VA 3.0 Physical Function Subscale (100 mm VAS), and Patient Global Assessment of Disease Status (PGAD) (100 mm VAS). The primary hypothesis was that etoricoxib 30 mg is non-inferior to celecoxib 200 mg as assessed by the primary endpoint (the non-inferiority margin was set at 10 mm VAS). Adverse events (AEs), laboratory parameters, and vital signs were monitored. There were 239 patients (89.5% female; mean age: 63.3 years) randomized to etoricoxib 30 mg (n = 120) and celecoxib 200 mg (n = 119). The differences (etoricoxib vs celecoxib) in least square (LS) mean change (95% CI) for WOMAC Pain, WOMAC Physical Function, and PGAD were -1.63 mm (-5.37, 2.10), -1.32 mm (-4.88, 2.23), and -1.09 mm (-5.48, 3.30), respectively. Drug-related clinical AEs occurred in 6.7% (etoricoxib) and 2.5% (celecoxib) of patients. This study was limited because it was not designed or powered to adequately capture and evaluate rare AEs associated with NSAID treatment. Etoricoxib 30 mg administered once daily in Korean patients with knee OA demonstrated non-inferior clinical efficacy to celecoxib 200 mg over 12 weeks of treatment as assessed by all primary and secondary outcomes. Etoricoxib 30 mg qd and celecoxib 200 mg qd were generally safe and well tolerated. NCT01554163.