Merck
  • Ribonucleotide reductase M2 is a promising molecular target for the treatment of oral squamous cell carcinoma.
Ribonucleotide reductase M2 is a promising molecular target for the treatment of oral squamous cell carcinoma.
International journal of oncology (2015-03-05)
Kazuki Iwamoto, Koh-Ichi Nakashiro, Hiroshi Tanaka, Norihiko Tokuzen, Hiroyuki Hamakawa
ABSTRACT
In our previous study, ribonucleotide reductase M2 (RRM2) was identified as a cancer-related gene commonly overexpressed in human oral squamous cell carcinoma (OSCC) cell lines. Herein, we attempted to determine whether targeting RRM2 may be a plausible therapeutic approach for the treatment of patients with OSCC. First, we examined the expression levels of RRM2 in human OSCC cell lines and tissues. Overexpression of RRM2 in OSCC was confirmed by western blot analysis. Subsequently, we investigated the effects of a synthetic small interfering RNA specific for RRM2 and gemcitabine (GEM), an inhibitor of RRM2 enzymatic activity, on the growth of human OSCC cell lines and primary cultured cells. Targeting RRM2 by RNA interference almost completely suppressed the expression of RRM2 and markedly suppressed the growth of both types of cells by >54.8%. GEM also reduced the growth rate of these cells by >83.0%. Finally, we evaluated the antitumor effects of GEM, cisplatin (CDDP), 5-fluorouracil (5-FU), and docetaxel (DOC) against OSCC cells using the collagen gel droplet embedded culture drug sensitivity test. OSCC cells were more sensitive to GEM and DOC than to CDDP and 5-FU, regardless of the expression level of RRM2 mRNA. These results suggested that RRM2 supported the growth of human OSCC cells and that targeting of RRM2, e.g., via GEM treatment, may be a promising therapeutic strategy for OSCC.
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Gemcitabine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
MISSION® esiRNA, targeting human RRM2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rrm2
USP
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
USP
Gemcitabine hydrochloride, United States Pharmacopeia (USP) Reference Standard
USP
Docetaxel, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Gemcitabine hydrochloride, ≥98% (HPLC)
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Anhydrous Docetaxel, European Pharmacopoeia (EP) Reference Standard
Supelco
5-Fluorouracil, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
21-Hydroxyprogesterone, ≥97% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Cisplatin, European Pharmacopoeia (EP) Reference Standard