Merck
  • Simultaneous determination of amino acid and monoamine neurotransmitters in PC12 cells and rats models of Parkinson's disease using a sensitizing derivatization reagent by UHPLC-MS/MS.
Simultaneous determination of amino acid and monoamine neurotransmitters in PC12 cells and rats models of Parkinson's disease using a sensitizing derivatization reagent by UHPLC-MS/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-05-30)
Xian-En Zhao, Shuyun Zhu, Hongmei Yang, Jinmao You, Fengrui Song, Zhiqiang Liu, Shuying Liu
ABSTRACT
Multi-analytes simultaneous monitoring of amino acid and monoamine neurotransmitters (NTs) has important scientific significance for their related pathology, physiology and drug screening. In this work, in virtue of a mass spectrometry sensitizing reagent 10-ethyl-acridone-3-sulfonyl chloride (EASC) as derivatization reagent, an Ultra High Performance Liquid Chromatography-Tandem Mass Spectrometry (UHPLC-MS/MS) method was developed and validated for simultaneous determination of six amino acid NTs, two monoamine ones and its one metabolite. The simple and rapid derivatization reaction was innovatively combined with plasma preparation by using EASC acetonitrile solution as protein precipitant. This interesting combination brought the advantages of speediness, simpleness and high-throughput in a cost-effective way. Under the optimized conditions, LODs (0.004-3.80nM) and LOQs (0.014-13.3nM) of EASC derivatized-NTs were calculated and found to be significantly lower than those of direct UHPLC-MS/MS detection about 11.5-275.0 and 14.4-371.4 times, respectively. Moreover, EASC derivatization significantly improved chromatographic resolution and matrix effect when compared with direct UPLC-MS/MS detection method without derivatization. Meanwhile, it also brought acceptable precision (3.0-13.0%, peak area CVs%), accuracy (86.4-112.9%), recovery (88.3-107.8%) and stability (3.8-8.5%, peak area CVs%) results. This method was successfully applied for the antiparkinsonian effect evaluation of levodopa and Ginsenoside Rg1 using PC12 cells and rats models by measuring multiple NTs. This provided a new method for the NTs related studies in the future.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Taurine, ≥98%, FG
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder
Sigma-Aldrich
D-Serine, ≥98% (TLC)
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
SAFC
Taurine
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)