Merck
  • Development and validation of a LC-MS/MS method for quantification of hetrombopag for pharmacokinetics study.
Development and validation of a LC-MS/MS method for quantification of hetrombopag for pharmacokinetics study.
SpringerPlus (2015-11-07)
Tao Chen, Zhonghua Chen, Suxing Zhang, Kezhi Zhang, Laiyou Wang
ABSTRACT
Hetrombopag as the derivative of ethylidene hydrazine carboxamide was recently developed into a novel patented non-peptide thrombopoietin mimetic and thrombopoietin receptor agonist to treat idiopathic thrombocytopenic purpura. To study the pharmacokinetics of hetrombopag, a highly sensitive, rapid and reliable liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was developed and validated for determination of hetrombopag in rat plasma. After protein precipitation extraction, the chromatography separation of analyte and internal standard named eltrombopag as an marketed analog of hetrombopag was performed on an Synergi Polar-RP column at the flow rate of 0.5 mL/min, and the determination was conducted on an API4000 triple quadrupole mass spectrometry in the multiple reaction monitoring mode using the respective [M+H](+) ions m/z 459.2 → 200.9 for hetrombopag and m/z 443.2 → 229.0 for IS. The lower limit of quantification was established to be 1 ng/mL, and the linear scope of standard curve was 1-1000 ng/mL. Both the precision (RSD%) and accuracy (RE%) were within the acceptable criterion of below 15 %. The validated method was successfully applied to quantify hetrombopag in the rat plasma and investigate the pharmacokinetics.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water