Merck
  • Microwave-assisted fabrication of chitosan-hydroxyapatite superporous hydrogel composites as bone scaffolds.
Microwave-assisted fabrication of chitosan-hydroxyapatite superporous hydrogel composites as bone scaffolds.
Journal of tissue engineering and regenerative medicine (2012-12-15)
Işıl Gerçek Beşkardeş, Tuğrul Tolga Demirtaş, Müge Dağlı Durukan, Menemşe Gümüşderelioğlu
ABSTRACT
In this study, a novel scaffold fabrication method was developed by combining microwave irradiation and gas foaming. Chitosan superporous hydrogels (SPHs) and chitosan-hydroxyapatite (HA) superporous hydrogel composites (SPHCs) were prepared by using this method in the presence of crosslinking agent, glyoxal, and a gas-blowing agent, NaHCO3. In order to examine the effect of HA on composite structure and cellular behaviour, two types of HA particles, i.e. spherical beads in 45-80 µm diameter and powder form, were used. While rapid heating with microwave irradiation enhances gas blowing, pH increment, which is accelerated by NaHCO3 decomposition, provides better crosslinking. Thus, interconnected and well-established macroporous hydrogels/hydrogel composites were produced easily and rapidly (~1 min). Cell culture studies, which were carried out under static and dynamic conditions with MC3T3-E1 pre-osteoblastic cells, indicated that chitosan-HA bead SPHCs supported cellular proliferation and osteoblastic differentiation better than chitosan SPHs and chitosan-HA powder SPHCs. In conclusion, simultaneous gas foaming and microwave crosslinking can be evaluated for the preparation of composite scaffolds which have superior properties for bone tissue engineering.
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Hydroxyapatite, powder, synthetic
Sigma-Aldrich
Glutaraldehyde solution, 25 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Glyoxal solution, BioReagent, ~40% in H2O (~8.8 M), Molecular Biology
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glyoxal solution, 40 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Calcium phosphate tribasic, BioReagent, suitable for plant cell culture, powder
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder