Merck
  • Enzymatic amplification-free nucleic acid hybridisation sensing on nanostructured thick-film electrodes by using covalently attached methylene blue.
Enzymatic amplification-free nucleic acid hybridisation sensing on nanostructured thick-film electrodes by using covalently attached methylene blue.
Talanta (2015-05-25)
Raquel García-González, Agustín Costa-García, M Teresa Fernández-Abedul
ABSTRACT
Amplification-free (referring to enzymatic amplification step) detection methodologies are increasing in biosensor development due to the need of faster and simpler protocols. However, for maintaining sensitivity without this step, highly detectable molecules or very sensitive detection techniques are required. The nanostructuration of transducer surfaces with carbon nanotubes (CNTs), gold nanoparticles (AuNPs) or both in nanohybrid configurations has been employed in this work for DNA hybridisation sensing purposes. Methylene blue (MB), covalently attached to single stranded DNA, (ssDNA) was incubated with a complementary sequence immobilized on nanostructured screen-printed electrodes (AuSPEs). Although CNTs can increase notoriously the signal of the marker, adsorptive properties should also be considered when bioassays are performed because non-specific adsorption (NSA) phenomena are magnified. In this work, strategies for decreasing NSA were thoroughly evaluated for the detection of Mycoplasma pneumoniae (MP) on CNTs-nanostructured screen-printed electrodes. Among them, the employ of UV-radiation or long incubation times (72h) allowed obtaining higher signals for the complementary strand with respect to the non-complementary one. The use of CNTs/AuNPs nanohybrids, together with the use of streptavidin-biotin (ST-B) interaction allows the higher differentiation (with a 3.5 ratio) in the genosensing of M. pneumoniae.
Sigma-Aldrich
Trizma® hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
6-Mercapto-1-hexanol, 99%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Trizma® hydrochloride, reagent grade, ≥99.0% (titration), crystalline
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Trizma® hydrochloride, BioPerformance Certified, suitable for cell culture, ≥99.0% (titration)
Sigma-Aldrich
Trizma® hydrochloride, pH 3.5-5.0 (0.5 M in H2O), BioXtra, ≥99.0% (titration)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Trizma® hydrochloride, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
1-Hexanethiol, 95%
Sigma-Aldrich
6-Mercapto-1-hexanol, 97%
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
1-Hexanethiol, 99% (GC)
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%