Merck
  • Global increase in O-linked N-acetylglucosamine modification promotes osteoblast differentiation.
Global increase in O-linked N-acetylglucosamine modification promotes osteoblast differentiation.
Experimental cell research (2015-08-25)
Takahiro Koyama, Kazuo Kamemura
ABSTRACT
The balance between bone formation and bone resorption is maintained by osteoblasts and osteoclasts, and an imbalance in this bone metabolism leads to osteoporosis. Here, we found that osteoblast differentiation in MC3T3-E1 cells is promoted by the inactivation of O-linked β-N-acetylglucosaminidase (O-GlcNAcase) and suppressed by the inactivation of O-GlcNAc transferase, as indicated by extracellular matrix calcification. The expression of osteogenic genes such as alp, ocn, and bsp during osteoblast differentiation was positively regulated in a O-GlcNAc glycosylation-dependent manner. Because it was confirmed that Ets1 and Runx2 are the two key transcription factors responsible for the expression of these osteogenic genes, their transcriptional activity might therefore be regulated by O-GlcNAc glycosylation. However, osteoclast differentiation of RAW264 cells, as indicated by the expression and activity of tartrate-resistant acid phosphatase, was unaffected by the inactivation of either O-GlcNAcase or O-GlcNAc transferase. Our findings suggest that an approach to manipulate O-GlcNAc glycosylation could be useful for developing the therapeutics for osteoporosis.
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
PUGNAc, ≥95% (HPLC)
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Msx2
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
MISSION® esiRNA, targeting human MSX2
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O