Merck
  • Antioxidant properties of carp (Cyprinus carpio L.) protein ex vivo and in vitro hydrolysates.
Antioxidant properties of carp (Cyprinus carpio L.) protein ex vivo and in vitro hydrolysates.
Food chemistry (2015-10-17)
Justyna Borawska, Małgorzata Darewicz, Gerd E Vegarud, Piotr Minkiewicz
ABSTRACT
The presence of specific peptides with antioxidant properties released during carp protein ex vivo and in vitro hydrolysis by human/porcine digestive enzymes, respectively, was examined. Based on the results of the in silico data analysis, antioxidant peptides were selected for subsequent identification in the digests/hydrolysates. Carp proteins were more resistant to hydrolysis by porcine enzymes than by human digestive juices. The sarcoplasmic proteins were hydrolyzed faster than the myofibrillar ones by both human/porcine enzymes. The in vitro myofibrillar protein hydrolysate showed the highest ABTS(+) scavenging activity (∼232.3 TEAC, μM Trolox/g), whereas the ex vivo hydrolysate of sarcoplasmic proteins showed the highest DPPH scavenging activity (∼88μM/g) and reducing power. Five antioxidant peptides were identified in carp protein ex vivo and in vitro hydrolysates: FIKK, HL, IY, PW, VY. The peptide HL from myofibrillar proteins was identified only in the ex vivo hydrolysate, whereas the peptide PW from sarcoplasmic proteins was identified only in the in vitro hydrolysate.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%