Merck
  • Determination of the Free Metal Ion Concentration Using AGNES Implemented with Environmentally Friendly Bismuth Film Electrodes.
Determination of the Free Metal Ion Concentration Using AGNES Implemented with Environmentally Friendly Bismuth Film Electrodes.
Analytical chemistry (2015-05-21)
Luciana S Rocha, Josep Galceran, Jaume Puy, José Paulo Pinheiro
ABSTRACT
Ex situ plated Bi film electrodes (Bi-FE) have been employed, for the first time, to measure the free concentration of Pb(II) in aqueous solutions using absence of gradients and Nernstian equilibrium stripping (AGNES) with stripping chronopotentiometry (SCP) quantification. The amount of deposited Pb°, below a certain threshold, follows a Nernstian relationship with the applied potential. This threshold can be interpreted as the frontier of transition from surface deposition to solid (bulk) formation of Pb°. AGNES with Bi-FE yielded a very good detection limit (3σ) for Pb(II) of 6.0 × 10(-9) M with an applied gain of 398 and a deposition time of 400 s. The ability of the Bi film electrode to perform speciation measurements was demonstrated for Pb(II)-PSS and Pb(II)-IDA systems. The measured values with the Bi-FE were in good agreement with the values obtained using the Hg film electrode and/or the values reported in the literature.
Sigma-Aldrich
4,4′-Azobis(4-cyanovaleric acid), ≥98.0% (T)
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Thioglycolic acid, 98%
Sigma-Aldrich
Thioglycolic acid, ≥97%
Sigma-Aldrich
Thioglycolic acid, ≥99%
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Thioglycolic acid solution, ~70 % (w/w) in H2O
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Ethanesulfonic acid, 95%