Merck
  • Genome-wide association study of corticobasal degeneration identifies risk variants shared with progressive supranuclear palsy.
Genome-wide association study of corticobasal degeneration identifies risk variants shared with progressive supranuclear palsy.
Nature communications (2015-06-17)
Naomi Kouri, Owen A Ross, Beth Dombroski, Curtis S Younkin, Daniel J Serie, Alexandra Soto-Ortolaza, Matthew Baker, Ni Cole A Finch, Hyejin Yoon, Jungsu Kim, Shinsuke Fujioka, Catriona A McLean, Bernardino Ghetti, Salvatore Spina, Laura B Cantwell, Martin R Farlow, Jordan Grafman, Edward D Huey, Mi Ryung Han, Sherry Beecher, Evan T Geller, Hans A Kretzschmar, Sigrun Roeber, Marla Gearing, Jorge L Juncos, Jean Paul G Vonsattel, Vivianna M Van Deerlin, Murray Grossman, Howard I Hurtig, Rachel G Gross, Steven E Arnold, John Q Trojanowski, Virginia M Lee, Gregor K Wenning, Charles L White, Günter U Höglinger, Ulrich Müller, Bernie Devlin, Lawrence I Golbe, Julia Crook, Joseph E Parisi, Bradley F Boeve, Keith A Josephs, Zbigniew K Wszolek, Ryan J Uitti, Neill R Graff-Radford, Irene Litvan, Steven G Younkin, Li-San Wang, Nilüfer Ertekin-Taner, Rosa Rademakers, Hakon Hakonarsen, Gerard D Schellenberg, Dennis W Dickson
ABSTRACT
Corticobasal degeneration (CBD) is a neurodegenerative disorder affecting movement and cognition, definitively diagnosed only at autopsy. Here, we conduct a genome-wide association study (GWAS) in CBD cases (n=152) and 3,311 controls, and 67 CBD cases and 439 controls in a replication stage. Associations with meta-analysis were 17q21 at MAPT (P=1.42 × 10(-12)), 8p12 at lnc-KIF13B-1, a long non-coding RNA (rs643472; P=3.41 × 10(-8)), and 2p22 at SOS1 (rs963731; P=1.76 × 10(-7)). Testing for association of CBD with top progressive supranuclear palsy (PSP) GWAS single-nucleotide polymorphisms (SNPs) identified associations at MOBP (3p22; rs1768208; P=2.07 × 10(-7)) and MAPT H1c (17q21; rs242557; P=7.91 × 10(-6)). We previously reported SNP/transcript level associations with rs8070723/MAPT, rs242557/MAPT, and rs1768208/MOBP and herein identified association with rs963731/SOS1. We identify new CBD susceptibility loci and show that CBD and PSP share a genetic risk factor other than MAPT at 3p22 MOBP (myelin-associated oligodendrocyte basic protein).
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, natural, 97%, FG