Merck
  • Copper(II) complexes of terminally free alloferon mutants containing two histidyl binding sites inside peptide chain structure and stability.
Copper(II) complexes of terminally free alloferon mutants containing two histidyl binding sites inside peptide chain structure and stability.
Dalton transactions (Cambridge, England : 2003) (2015-11-14)
Agnieszka Kadej, Mariola Kuczer, Teresa Kowalik-Jankowska
ABSTRACT
Mononuclear and polynuclear copper(II) complexes of alloferon 1 with point mutations, H1A/H12A H2N-A(1)GVSGH(6)GQH(9)GVA(12)G-COOH, H1A/H9A H2N-A(1)GVSGH(6)GQA(9)GVH(12)G-COOH, and H1A/H6A H2N-A(1)GVSGA(6)GQH(9)GVH(12)G-COOH, have been studied by potentiometric, UV-visible, CD, and EPR spectroscopy, and mass spectrometry (MS) methods. Complete complex speciation at different metal-to-ligand molar ratios ranging from 1 : 1 to 3 : 1 was obtained. Over a wide 6-8 pH range, including physiological pH 7.4, and a 1 : 1 metal-to-ligand molar ratio, the peptides studied formed a CuH-1L complex with the 4N{NH2,N(-),2NIm} coordination mode. The presence of the 4N binding site for the CuH-1L complexes prevented the deprotonation and coordination of the second amide nitrogen atom to copper(II) ions (pK-1/-2 7.83-8.07) compared to that of pentaGly (6.81). The amine nitrogen donor and two imidazole nitrogen atoms (H(6)H(9), H(6)H(12) and H(9)H(12)) can be considered to be independent metal-binding sites in the species formed. As a consequence, di- and trinuclear complexes for the metal-to-ligand 2 : 1 and 3 : 1 molar ratios dominate in the solution, respectively. For the Cu(II)-H1A/H9A and Cu(II)-H1A/H12A systems, the Cu3H-9L complexes are likely formed by the coordination of amide nitrogen atoms towards C-termini with ring sizes (7,5,5).
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Potassium nitrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium nitrate, puriss., meets analytical specification of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%
Sigma-Aldrich
Potassium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥65%
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets