Merck
miR-126 inhibits papillary thyroid carcinoma growth by targeting LRP6.
Oncology reports (2015-08-05)
Qiang Wen, Jie Zhao, Lin Bai, Tongtong Wang, Haishan Zhang, Qingjie Ma
ABSTRACT
microRNA-126 (miR-126) has been reported to play tumor suppressor roles in various types of cancers. Although it has been reported that miR-126 expression is downregulated in papillary thyroid carcinoma (PTC), the precise role and underlying molecular mechanism of miR-126 in PTC remains unclear. Therefore, the aims of the present study were to investigate the role and potential mechanism of miR-126 in tumorigenicity of PTC in vivo and in vitro. We observed that the miR-126 expression level was significantly downregulated in PTC tissue and PTC cell lines, the aberrant expression of miR-126 was correlated with lymph node metastasis, tumor size and TNM stage. We also showed that restoration of miR-126 in PTC cells inhibited cell proliferation, colony formations, migration and invasion, promoted cell apoptosis and cell cycle arrest at G1 stage in vitro, as well as inhibited tumor growth and decreased tumor volume and weight in vivo. Furthermore, low-density lipoprotein receptor‑related protein 6 (LRP6), a regulator of the Wnt/β‑catenin signaling cascade, was identified as a crucial target gene of miR-126. Overexpression of miR-126 inhibited LP6 expression on mRNA and protein levels, and deactivate Wnt/β-catenin signaling pathway. These results suggested that miR-126 functions as a tumor-suppressive miRNA by targeting LRP6 regulating Wnt/β-catenin signaling pathway and represents a therapeutic target for PTC.
Sigma-Aldrich
meso-2,3-Dimercaptosuccinic acid, ~98%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfide, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfide, anhydrous, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfide, ≥99%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfide, natural, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma