Merck
  • Validation of an analytical method for human plasma free amino acids by high-performance liquid chromatography ionization mass spectrometry using automated precolumn derivatization.
Validation of an analytical method for human plasma free amino acids by high-performance liquid chromatography ionization mass spectrometry using automated precolumn derivatization.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-07-18)
Hiroo Yoshida, Kazuhiro Kondo, Hiroyuki Yamamoto, Naoko Kageyama, Shin-Ichi Ozawa, Kazutaka Shimbo, Takahiko Muramatsu, Akira Imaizumi, Toshimi Mizukoshi, Junichi Masuda, Daisuke Nakayama, Yoshihiro Hayakawa, Kyoko Watanabe, Kazuo Mukaibatake, Hiroshi Miyano
ABSTRACT
The analysis of human plasma free amino acids is important for diagnosing the health of individuals, because their concentrations are known to vary with various diseases. The development of valid, reliable, and high-throughput analytical methods for amino acids analysis is an essential requirement in clinical applications. In the present study, we have developed an automated precolumn derivatization amino acid analytical method based on high-performance liquid chromatography/electrospray ionization mass spectrometry (so-called UF-Amino Station). This method enabled the separation of at least 38 types of physiological amino acids within 8min, and the interval time between injections was 12min. We also validated this method for 21 major types of free amino acids in human plasma samples. The results of the specificity, linearity, accuracy, repeatability, intermediate precision, reproducibility, limits of detections, lower limits of quantification, carry over, and sample solution stability were sufficient to allow for the measurement of amino acids in human plasma samples. Our developed method should be suitable for use in clinical fields.
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
DL-2-Aminobutyric acid, ReagentPlus®, 99%
Sigma-Aldrich
L-Asparagine, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
L-Asparagine, ≥98% (HPLC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
L-Cystine, Wacker Chemie AG, ≥98.5%
Sigma-Aldrich
Ninhydrin, suitable for amino acid detection
Sigma-Aldrich
Ninhydrin, ACS reagent
SAFC
L-Cystine
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%