Merck
  • Radical Cationic Pathway for the Decay of Ionized Glyme Molecules in Liquid Solution.
Radical Cationic Pathway for the Decay of Ionized Glyme Molecules in Liquid Solution.
The journal of physical chemistry. B (2015-10-17)
Konstantin S Taletskiy, Vsevolod I Borovkov, Lyudmila N Schegoleva, Irina V Beregovaya, Andrey I Taratayko, Yuriy N Molin
ABSTRACT
Chemical stability of primary radical cations (RCs) generated in irradiated matter determines substantially the radiation resistance of organic materials. Transformations of the RCs of the glyme molecules, R(-O-CH2-CH2-)nO-R (R = CH3, n = 1-4) has been studied on the nanosecond time scale by measuring the magnetic field effects in the recombination fluorescence from irradiated liquid solutions of the glymes. In all cases, the RCs observed were different from that expected for the primary ones and revealed very similar hyperfine couplings independent of the poly(ethylene oxide) chain length and of the substitution of terminal methyl groups by C2H5 or CH2CH2Cl, as has been shown with diglyme as an example. Quantum chemical analysis of possible chemical transformations for the monoglyme RC as a model system allowed us to discover the reaction pathway yielding the methyl vinyl ether RC. The pathway involves intramolecular proton transfer followed by C-O bond cleavage. Only one (-O-CH2-CH2-O-) fragment is involved in this transformation, which is nearly barrierless due to the catalytic effect of adjacent glyme molecules. The rapid formation of the methyl vinyl ether RC in the irradiated monoglyme was confirmed by the numerical simulation of the experimental curves of the time-resolved magnetic field effect. These findings suggest that the R'-O-CH═CH2(•+) formation is a typical decay pathway for the primary RCs in irradiated liquid glymes.
Sigma-Aldrich
1,2-Dimethoxyethane, anhydrous, 99.5%, inhibitor-free
Sigma-Aldrich
Decane, anhydrous, ≥99%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Decane, ≥95%
Sigma-Aldrich
Diethylene glycol dimethyl ether, anhydrous, 99.5%
Sigma-Aldrich
Tetraethylene glycol dimethyl ether, ≥99%
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Decane, ReagentPlus®, ≥99%
Sigma-Aldrich
Diethylene glycol dimethyl ether, ReagentPlus®, 99%
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
1,2-Dimethoxyethane, ReagentPlus®, ≥99%, inhibitor-free
Sigma-Aldrich
Diethyl ether, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Triethylene glycol dimethyl ether, ReagentPlus®, 99%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%