Merck
pH-responsive scaffolds generate a pro-healing response.
Biomaterials (2015-05-10)
Jin-Oh You, Marjan Rafat, Dariela Almeda, Natalia Maldonado, Peng Guo, Christoph S Nabzdyk, Maggie Chun, Frank W LoGerfo, John W Hutchinson, Leena K Pradhan-Nabzdyk, Debra T Auguste
ABSTRACT
A principal challenge in wound healing is a lack of cell recruitment, cell infiltration, and vascularization, which occurs in the absence of temporal and spatial cues. We hypothesized that a scaffold that expands due to local changes in pH may alter oxygen and nutrient transport and the local cell density, leading to enhanced cell deposition and survival. In this study, we present a pH-responsive scaffold that increases oxygen transport, as confirmed by our finite element model analysis, and cell proliferation relative to a non-responsive scaffold. In vivo, responsive scaffolds induce a pro-healing gene expression profile indicative of enhanced angiogenesis, granulation tissue formation, and tissue remodeling. Scaffolds that stretch in response to their environment may be a hallmark for tissue regeneration.
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Hydroxyethyl methacrylate
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
2-Hydroxyethyl methacrylate, ≥99%, contains ≤50 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
2-Hydroxyethyl methacrylate, contains ≤250 ppm monomethyl ether hydroquinone as inhibitor, 97%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Tetraethylene glycol dimethacrylate, technical, ≥90% (GC)
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer