Merck
  • Fast preparation of a highly efficient organic monolith via photo-initiated thiol-ene click polymerization for capillary liquid chromatography.
Fast preparation of a highly efficient organic monolith via photo-initiated thiol-ene click polymerization for capillary liquid chromatography.
Journal of chromatography. A (2015-04-09)
Lianfang Chen, Junjie Ou, Zhongshan Liu, Hui Lin, Hongwei Wang, Jing Dong, Hanfa Zou
ABSTRACT
A novel organic monolith was firstly prepared in a UV-transparent fused-silica capillary by a single-step approach via photo-initiated thiol-ene click polymerization reaction of 1,2,4-trivinylcyclohexane (TVCH) and pentaerythriol tetra(3-mercaptopropionate) (4SH) within 10min. The effects of both composition of prepolymerization solution and polymerization time on the morphology and permeability of monolithic column were investigated in detail. Then, the optimal condition was acquired to fabricate a homogeneous and permeable organic monolith. The chemical groups of the monolithic column were confirmed by Fourier transform infrared spectroscopy (FT-IR). The SEM graphs showed the organic monolith possessed a uniform porous structure, which promotes the highest column efficiency of ∼133,000 plates per meter for alkylbenzenes at the linear velocity of 0.65mm/s in reversed-phase liquid chromatography. Finally, the organic monolithic column was further applied for separation of basic compounds, pesticides and EPA610, indicating satisfactory separation ability.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Vinyltrimethoxysilane, 97%
Sigma-Aldrich
1,6-Hexanedithiol, 96%
Sigma-Aldrich
1,6-Hexanedithiol, ≥97%, FG
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Vinyltrimethoxysilane, 98%
Sigma-Aldrich
1,1,1-Tris(hydroxymethyl)propane, dist., ≥98.0% (GC)
Sigma-Aldrich
1,1,1-Tris(hydroxymethyl)propane, 97%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)