Merck
  • Tissue Transglutaminase, Not Lysyl Oxidase, Dominates Early Calcium-Dependent Remodeling of Fibroblast-Populated Collagen Lattices.
Tissue Transglutaminase, Not Lysyl Oxidase, Dominates Early Calcium-Dependent Remodeling of Fibroblast-Populated Collagen Lattices.
Cells, tissues, organs (2015-05-01)
D D Simon, L E Niklason, J D Humphrey
ABSTRACT
Cell-populated collagen gels have provided significant insight into the cellular contractile mechanisms and cell-matrix interactions that are necessary for compacting and remodeling extant matrix. Nevertheless, little research has been devoted towards determining how cells entrench these deformations that contribute to establishing a preferred mechanical state. To this end, we examined the roles of two covalent matrix cross-linkers, i.e. tissue transglutaminase and lysyl oxidase, during global remodeling of the free-floating fibroblast-populated collagen lattice. Inhibition of tissue transglutaminase resulted in a reduced rate of compaction compared to controls during early remodeling (up to 2 days). In contrast, inhibition of lysyl oxidase did not alter the early compaction of these lattices, but it reduced the compaction after 2 days of culture. Acute inhibition of different contractile mechanisms suggested further that calcium-dependent contractility may have dominated during the initial remodeling of the collagen lattice before giving way to calcium-independent contractility at later times. In summary, these findings suggest that early remodeling of the free-floating collagen lattice is facilitated by calcium-dependent cell contraction while entrenchment is dominated by a tissue transglutaminase-mediated cross-linking of the extant matrix. As remodeling continues, however, lysyl oxidase increases its contribution, perhaps by consolidating de novo collagen fibrils into fibers to continue the remodeling while the cells transition to a more sustained, calcium-independent contractility. These results promise to influence future tissue engineering studies as well as computational simulations aimed at understanding matrix remodeling in complex in vivo situations.
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
SAFC
Glycine
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%