Merck
  • Mechanisms of cholesterol and saturated fatty acid lowering by Quillaja saponaria extract, studied by in vitro digestion model.
Mechanisms of cholesterol and saturated fatty acid lowering by Quillaja saponaria extract, studied by in vitro digestion model.
Food & function (2015-03-17)
Liliya Vinarova, Zahari Vinarov, Borislava Damyanova, Slavka Tcholakova, Nikolai Denkov, Simeon Stoyanov
ABSTRACT
Quillaja saponin extracts are known to reduce plasma cholesterol levels in humans. Here we study the mechanism of this effect with Quillaja Dry saponin extract (QD). In vitro model of triglyceride lipolysis is used to quantify the effect of QD on the solubilization of cholesterol and of the lipolysis products (fatty acids and monoglycerides) in the dietary mixed micelles (DMM). We found that QD extract decreases significantly both the cholesterol (from 80% to 20%) and saturated fatty acids (SFA, from 70% to 10%) solubilised in DMM. Series of dedicated experiments prove that QD may act by two mechanisms: (1) direct precipitation of cholesterol and (2) displacement of cholesterol from the DMM. Both mechanisms lead to increased cholesterol precipitation and, thus, render cholesterol bio-inaccessible. We prove also that the saponin molecules are not the active component of QD, because highly purified Quillaja extract with very similar saponin composition does not exhibit cholesterol-lowering or SFA-lowering effect. The effect of QD extract on cholesterol solubilisation is most probably caused by the high-molecular weight polyphenol molecules, present in this extract. The reduced SFA solubilisation is caused by Ca(2+) ions of relatively high concentration (1.25 wt%), also present in QD extract, which precipitate the fatty acids into calcium soaps.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Myristic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Dipalmitin, ≥99.0%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium glycodeoxycholate, BioXtra, ≥97% (HPLC)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Myristic acid, natural, ≥98.5%, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG