Merck
  • Synthesis and evaluation of the structural and physicochemical properties of carboxymethyl pregelatinized starch as a pharmaceutical excipient.
Synthesis and evaluation of the structural and physicochemical properties of carboxymethyl pregelatinized starch as a pharmaceutical excipient.
Saudi pharmaceutical journal : SPJ : the official publication of the Saudi Pharmaceutical Society (2015-12-25)
Sonia Lefnaoui, Nadji Moulai-Mostefa
ABSTRACT
A pregelatinized starch (PGS) was derivatized with sodium chloroacetate (SCA) in alcoholic medium under alkaline condition to produce carboxymethyl pregelatinized starch (CMPGS) with various degrees of substitution (DS). Influence of the molar ratio of SCA to the glucopyranose units (SCA/GU), reaction time, temperature and the amount of sodium hydroxide on the degree of substitution (DS) and the reaction efficiency (RE) was studied. An optimal concentration of 30% of NaOH, for a reaction time of 1 h at 50 °C and molar ratio (SCA/GU) equal to 1.0, yielded an optimal DS of 0.55 and a RE of 55%. SEM micrographs revealed that the carboxymethylation assigned the structural arrangement of CMPGS and caused the granular disintegration. Wide angle diffraction X-ray (XRD) showed that the crystallinity of starch was obviously varied after carboxymethylation. New bands in FTIR spectra at 1417 and 1603 cm(-1) indicated the presence of carboxymethyl groups. The solubility and viscosity of CMPGS increased with an increase in the degree of modification. In order to investigate the influence of DS on physical and drug release properties, CMPGS obtained with DS in the range of 0.12-0.55 was evaluated as tablet excipient for sustained drug release. Dissolution tests performed in phosphate buffer (pH 6.8), with Ibuprofen as drug model (25% loading) showed that CMPGS seems suitable to be used as sustained release excipient since the drug release was driven over a period up to 8 h. The in vitro release kinetics studies revealed that all formulations fit well with Korsmeyer-Peppas model and the mechanism of drug release is non-Fickian diffusion.
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder