Merck
  • In vitro immunosuppressive and cytotoxic activities of Tripterygium wilfordii extract.
In vitro immunosuppressive and cytotoxic activities of Tripterygium wilfordii extract.
Drug and chemical toxicology (2014-05-23)
Yu He, Senlin Shi, Ruping Zhang, Wei Shen, Jue Tu, Zhishan Ding, Yongsheng Fan
ABSTRACT
Tripterygium wilfordii Hook. f. (TW) is a traditional herbal medicine which has been widely used for the treatment of rheumatoid arthritis and other autoimmune diseases. However, adverse reactions of TW such as hepatotoxicity and nephrotoxicity have been frequently reported in clinic. With the aim to evaluate the potency and toxicity of TW, we collected eleven batches of TW from different localities across Chinese mainland, and investigated the inhibition of their methanol extracts on the proliferation of mouse spleen lymphocytes, normal human hepatocyte (L-02) cells and African green monkey kidney (COS-7) cells. TW extracts with three different concentrations were designed as the experimental groups. Our present findings provided consistent evidence that TW had significant concentration-dependent inhibitory action on lymphocytes, L-02 and COS-7 cells. At the concentrations of 0.75 and 1.5 mg/mL, most TW groups showed statistically significant inhibition of lymphocyte proliferation when compared with the control group (p < 0.01), and the inhibition of TW extract on lymphocytes was almost equal to 1.0 mg/mL aspirin (p > 0.05). In most test groups, significant toxicities were shown on L-02 cells at 0.6 and 3.0 mg/mL (p < 0.01), and on COS-7 cells at 3.0 mg/mL (p < 0.01). At 3.0 mg/mL, almost all TW groups exerted obvious toxicities toward L-02 and COS-7 cells which were equal to or even higher than 1.0 mg/mL aspirin. In view of these results, further studies are needed to elucidate the relations among the effective component, curative effect and toxicity of TW to ensure its effectiveness and safety for human consumption.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture