Merck
  • MFAP5 is related to obesity-associated adipose tissue and extracellular matrix remodeling and inflammation.
MFAP5 is related to obesity-associated adipose tissue and extracellular matrix remodeling and inflammation.
Obesity (Silver Spring, Md.) (2015-06-09)
Maija Vaittinen, Marjukka Kolehmainen, Mikael Rydén, Matti Eskelinen, Martin Wabitsch, Jussi Pihlajamäki, Matti Uusitupa, Leena Pulkkinen
ABSTRACT
Low-grade inflammation is involved in adipose tissue (AT) and extracellular matrix (ECM) remodeling and induces deposition of ECM proteins in AT. We have previously shown that MFAP5 (microfibrillar-associated protein 5) expression decreases in AT after weight loss. The aim of this study was to investigate MFAP5 localization in human AT and gene expression in adipocytes and the role of MFAP5 in adipocyte metabolism and inflammation. MFAP5 protein localization and gene expression were studied with immunohistochemistry and quantitative reverse transcriptase PCR (RT-qPCR) in human subcutaneous AT and cultured Simpson-Golabi-Behmel syndrome (SGBS) adipocytes, respectively. The effect of MFAP5 knock-down by siRNA on gene expression and insulin action was examined with RT-qPCR, western blot, and insulin-stimulated glucose uptake. The effect of different cytokines on MFAP5 gene and protein expression was investigated in cultured human SGBS preadipocytes. MFAP5 protein was highly expressed in AT, and gene expression decreased during adipocyte differentiation in SGBS cells. Treatment of preadipocytes with TNFα and TGFβ1 increased MFAP5 gene and protein expression. Furthermore, MFAP5 knock-down decreased the expression of genes involved in inflammation. Our results demonstrate that factors involving low-grade inflammation modulate MFAP5 expression and that the modified expression of MFAP5 may further regulate AT inflammation.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
MISSION® esiRNA, targeting human MFAP5
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mfap5
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)