Merck
  • Comprehensive thin layer chromatography×gas chromatography using headspace sampling modulation-A case study on fatty acid composition analysis.
Comprehensive thin layer chromatography×gas chromatography using headspace sampling modulation-A case study on fatty acid composition analysis.
Journal of chromatography. A (2015-06-24)
Mitja Križman, Alenka Pušar
ABSTRACT
The recently developed comprehensive TLC×GC technique using headspace sampling is presented. The main advantage of this approach, as demonstrated in lipid analysis, is the possibility to include a transesterification step of glycerides into fatty acid methyl ester derivatives (FAME) because no particular constraints in terms of operational time between TLC and GC are present. Besides being a relatively low-cost solution, TLC×GC by means of headspace sampling provides many benefits in terms of flexibility of separation conditions and modulation sampling width. The technique provides over two orders of magnitude of linear range with TLC sample loads of about 1mg with good reproducibility and accuracy, as demonstrated by multiple headspace extraction (MHE) tests. The technique is a viable alternative to the established but more expensive HPLC×GC technique. The useful range of TLC×GC in terms of analyte volatility can be further extended with a future development of devices based on thermal desorption.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Diisopropyl ether, anhydrous, 99%, contains either BHT or hydroquinone as stabilizer
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis