Merck
  • Fe65 Suppresses Breast Cancer Cell Migration and Invasion through Tip60 Mediated Cortactin Acetylation.
Fe65 Suppresses Breast Cancer Cell Migration and Invasion through Tip60 Mediated Cortactin Acetylation.
Scientific reports (2015-07-15)
Yuefeng Sun, Jianwei Sun, Panida Lungchukiet, Waise Quarni, Shengyu Yang, Xiaohong Zhang, Wenlong Bai
ABSTRACT
Fe65 is a brain-enriched adaptor protein known for its role in the action of the Aβ amyloid precursor protein in neuronal cells and Alzheimer's disease, but little is known about its functions in cancer cells. The present study documents for the first time a role of Fe65 in suppressing breast cancer cell migration and invasion. Mechanistic studies suggest that the suppression is mediated through its phosphotyrosine binding domain 1 that mediates the recruitment of Tip60 to cortactin to stimulate its acetylation. The studies identify the Tip60 acetyltransferase as a cytoplasmic drug target for the therapeutic intervention of metastatic breast cancers.
Sigma-Aldrich
MISSION® esiRNA, targeting human APBB1
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Apbb1
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.