Merck
  • IL-21 induces antiviral microRNA-29 in CD4 T cells to limit HIV-1 infection.
IL-21 induces antiviral microRNA-29 in CD4 T cells to limit HIV-1 infection.
Nature communications (2015-06-26)
Stanley Adoro, Juan R Cubillos-Ruiz, Xi Chen, Maud Deruaz, Vladimir D Vrbanac, Minkyung Song, Suna Park, Thomas T Murooka, Timothy E Dudek, Andrew D Luster, Andrew M Tager, Hendrik Streeck, Brittany Bowman, Bruce D Walker, Douglas S Kwon, Vanja Lazarevic, Laurie H Glimcher
ABSTRACT
Initial events after exposure determine HIV-1 disease progression, underscoring a critical need to understand host mechanisms that interfere with initial viral replication. Although associated with chronic HIV-1 control, it is not known whether interleukin-21 (IL-21) contributes to early HIV-1 immunity. Here we take advantage of tractable primary human lymphoid organ aggregate cultures to show that IL-21 directly suppresses HIV-1 replication, and identify microRNA-29 (miR-29) as an antiviral factor induced by IL-21 in CD4 T cells. IL-21 promotes transcription of all miR-29 species through STAT3, whose binding to putative regulatory regions within the MIR29 gene is enriched by IL-21 signalling. Notably, exogenous IL-21 limits early HIV-1 infection in humanized mice, and lower viremia in vivo is associated with higher miR-29 expression. Together, these findings reveal a novel antiviral IL-21-miR-29 axis that promotes CD4 T-cell-intrinsic resistance to HIV-1 infection, and suggest a role for IL-21 in initial HIV-1 control in vivo.
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
SAFC
Glycine
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)