Merck
  • Effects of 4-hexylresorcinol on the phenoloxidase from Hyphantria cunea (Lepidoptera: Arctiidae): In vivo and in vitro studies.
Effects of 4-hexylresorcinol on the phenoloxidase from Hyphantria cunea (Lepidoptera: Arctiidae): In vivo and in vitro studies.
Insect science (2014-07-06)
Mahboobeh Sharifi, Mohammad Ghadamyari, Reza H Sajedi, Nosrat O Mahmoodi
ABSTRACT
Insecticidal effects of 4-hexylresorcinol, a phenoloxidase (PO) inhibitor, were determined on Hyphantria cunea (Drury) under laboratory conditions. The LC50 for the 15-d-old larvae was estimated to be 2.95 g/L after 96 h exposure. The activities of glutathione S-transferase (GST) and PO showed a decrease in larvae treated with 4-hexylresorcinol, and the IC50 of GST and PO were estimated to be 0.8 and 0.43 g/L, respectively, 24 h after treatment. The PO from the hemolymph of fall webworm was purified by ammonium sulfate precipitation, gel-filtration, and ion-exchange chromatography, and then enzymatic characteristics and the mechanism of inhibition were determined using L-dihydroxyphenylalanine (L-DOPA) as the substrate. The purified PO showed a single band on SDS-PAGE with a molecular weight of about 70 kDa. The optimum pH for PO activity was observed at pH 7.0, optimum temperature was found to be 45 °C, and PO activity was strongly inhibited by Zn(2+) . IC50 values were estimated to be 8.2, 19.14, and 24.04 μmol/L for 4-hexylresorsinol, kojic acid, and quercetin, respectively. The inhibitory potencies (i.e., I50 of each compound/I50 of 4-hexylresorcinol) of kojic acid and quercetin on H. cunea PO were estimated to be 1.87 and 2.89, respectively. 4-hexylresorcinol was determined to be a competitive inhibitor, and kojic acid and quercetin were determined to be mixed inhibitors. PO is one of the most important enzymes in an insect's immune system, and the use of PO inhibitors seems to be a promising approach for pest control due to their potential safety for humans.
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
2-(Diethylamino)ethanol, ≥99.5%
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Kojic acid
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, 97%
Sigma-Aldrich
4-Hexylresorcinol, 98%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Pyrogallol, ACS reagent
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%