Merck
  • The seed of the Amazonian fruit Couepia bracteosa exhibits higher scavenging capacity against ROS and RNS than its shell and pulp extracts.
The seed of the Amazonian fruit Couepia bracteosa exhibits higher scavenging capacity against ROS and RNS than its shell and pulp extracts.
Food & function (2015-07-28)
Alessandra Berto, Alessandra Braga Ribeiro, Enrique Sentandreu, Nilson Evelázio de Souza, Adriana Zerlotti Mercadante, Renan Campos Chisté, Eduarda Fernandes
ABSTRACT
Among the large number of scientifically unstudied fruits from the Amazonia biome, Couepia bracteosa acts as an interesting source of bioactive compounds, such as phenolic compounds and carotenoids, which may be used for protecting human health against oxidative damage. For the first time, the phenolic compounds and carotenoids in extracts obtained from the pulp, shell and seeds of C. bracteosa fruits are reported, as well as their in vitro scavenging capacities against some reactive oxygen species (ROS) and reactive nitrogen species (RNS). The shell extract presented the highest phenolic compound and carotenoid contents (5540 and 328 μg per g extract, dry basis, respectively), followed by the pulp and seed extracts. The major phenolic compound was acacetin sulphate (one methoxy and two OH groups) (62%) in the shells; however, only seeds presented apigenin sulphate (three OH groups), in which it was the major compound (44%). The high content of apigenin sulphate may explain why the seed extract had the highest scavenging efficiency against all tested ROS/RNS among the studied extracts. Regarding carotenoids, all-trans-neochrome (17%) and all-trans-β-carotene (16%) were the major carotenoids in the pulp extracts, while all-trans-lutein (44%) was the most prevalent in the shell extracts and all-trans-α-carotene (32%) and all-trans-β-carotene (29%) were the major ones in the seed extracts.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
NOC-5, ≥90%
Sigma-Aldrich
Phenazine methosulfate
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Phenazine, 98%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Acacetin, ≥97.0% (HPLC)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Dihydrorhodamine 123, ≥95%
Sigma-Aldrich
Xanthophyll, from marigold
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)