Merck
  • Enantiomers of 3-methylspermidine selectively modulate deoxyhypusine synthesis and reveal important determinants for spermidine transport.
Enantiomers of 3-methylspermidine selectively modulate deoxyhypusine synthesis and reveal important determinants for spermidine transport.
ACS chemical biology (2015-02-18)
Mervi T Hyvönen, Maxim Khomutov, Marine Petit, Janne Weisell, Sergey N Kochetkov, Leena Alhonen, Jouko Vepsäläinen, Alex R Khomutov, Tuomo A Keinänen
ABSTRACT
Eukaryotic translation initiation factor 5A (eIF5A) is essential for cell proliferation, becoming functionally active only after post-translational conversion of a specific Lys to hypusine [N(ε)-(4-amino-2-hydroxybutyl)lysine]. Deoxyhypusine synthase (DHS) is the rate-limiting enzyme of this two-step process, and the polyamine spermidine is the only natural donor of the butylamine group for this reaction, which is very conserved-hypusine biosynthesis suffers last when the intracellular spermidine pool is depleted. DHS has a very strict substrate specificity, and only a few spermidine analogs are substrates of the enzyme and can support long-term growth of spermidine-depleted cells. Herein, we compared the biological properties of earlier unknown enantiomers of 3-methylspermidine (3-MeSpd) in deoxyhypusine synthesis, in supporting cell growth and in polyamine transport. Long-term treatment of DU145 cells with α-difluoromethylornithine (inhibitor of polyamine biosynthesis) and (R)-3-MeSpd did not cause depletion of hypusinated eIF5A, and the cells were still able to grow, whereas the combination of α-difluoromethylornithine with a racemate or (S)-3-MeSpd caused cessation of cell growth. Noticeably, DHS preferred the (R)- over the (S)-enantiomer as a substrate. (R)-3-MeSpd competed with [(14)C]-labeled spermidine for cellular uptake less efficiently than the (S)-3-MeSpd (Ki = 141 μM vs 19 μM, respectively). The cells treated with racemic 3-MeSpd accumulated intracellularly mainly (S)-3-MeSpd, but not DHS substrate (R)-3-MeSpd, explaining the inability of the racemate to support long-term growth. The distinct properties of 3-MeSpd enantiomers can be exploited in designing polyamine uptake inhibitors, facilitating drug delivery and modulating deoxyhypusine synthesis.
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Ammonium sulfate, Molecular Biology, ≥99.0%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 16O, 99.99 atom % 14N
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%