Merck
  • IgM exacerbates glomerular disease progression in complement-induced glomerulopathy.
IgM exacerbates glomerular disease progression in complement-induced glomerulopathy.
Kidney international (2015-05-07)
Sarah E Panzer, Jennifer Laskowski, Brandon Renner, Liudmila Kulik, Danica Ljubanovic, Kendra M Huber, Weixiong Zhong, Matthew C Pickering, V Michael Holers, Joshua M Thurman
ABSTRACT
Although glomerular immunoglobulin M (IgM) deposition occurs in a variety of glomerular diseases, the mechanism of deposition and its clinical significance remain controversial. Some have theorized IgM becomes passively trapped in areas of glomerulosclerosis. However, recent studies found that IgM specifically binds damaged glomeruli. Therefore, we tested whether natural IgM binds to neo-epitopes exposed after insults to the glomerulus and exacerbates disease in mice deficient in the complement regulatory protein factor H; a model of non-sclerotic and nonimmune-complex glomerular disease. Immunofluorescence microscopy demonstrated mesangial and capillary loop deposition of IgM, whereas ultrastructural analysis found IgM deposition on endothelial cells and subendothelial areas. Factor H-deficient mice lacking B cells were protected from renal damage, as evidenced by milder histologic lesions on light and electron microscopy. IgM, but not IgG, from wild-type mice bound to cultured murine mesangial cells. Furthermore, injection of purified IgM into mice lacking B cells bound within the glomeruli and induced proteinuria. A monoclonal natural IgM-recognizing phospholipids also bound to glomeruli in vivo and induced albuminuria. Thus, our results indicate specific IgM antibodies bind to glomerular epitopes and that IgM contributes to the progression of glomerular damage in this mouse model of non-sclerotic glomerular disease.
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)