Merck
  • Validation of a liquid chromatography-high-resolution mass spectrometry method for the analysis of ceftiofur in poultry muscle, kidneys and plasma: A unique accuracy profile for each and every matrix.
Validation of a liquid chromatography-high-resolution mass spectrometry method for the analysis of ceftiofur in poultry muscle, kidneys and plasma: A unique accuracy profile for each and every matrix.
Journal of chromatography. A (2015-07-15)
Sophie Mompelat, Marie-Pierre Fourmond, Michel Laurentie, Eric Verdon, Dominique Hurtaud-Pessel, Jean-Pierre Abjean
ABSTRACT
A robust, selective and specific liquid chromatography-high resolution mass spectrometry (LC-HRMS) method was developed for the quantification of total residues of ceftiofur, an antibiotic belonging to the 3rd generation cephalosporins in plasma, muscle and kidney of poultry. Ceftiofur and conjugates in samples were firstly hydrolyzed with dithioerythritol into desfuroylceftiofur, which was then stabilized by derivatization with iodoacetamide into desfuroylceftiofur acetamide. Sample were then submitted to a solid phase extraction followed the accurate mass analysis of desfuroylceftiofur acetamide by LC-HRMS in full scan mode using a linear trap quadrupole (LTQ)-Orbitrap mass spectrometer with a resolving power 60,000 full width at half maximum (FWHM). The method was fully validated over a dosing range between 100 and 2000 μg kg(-1) (or μg L(-1)) using the total error approach. Accuracy profiles a graphical decision-making tool were built by computing results of validation procedure with acceptance limits set at ±60%, and β-expectation tolerance intervals, i.e. the interval assuming to contain a β % of future measurements (β=90% in this study). Total measurement error including trueness, repeatability and intermediate precision were evaluated. Relative bias of trueness was never exceeding the threshold of 6% in all matrices at all level of concentration. The mean relative standard deviation for repeatability was lower than 16% at all levels of concentration for all matrices; the mean relative standard deviation for intermediate precision was lower than 25% at all levels of concentration for all matrices. This validation approach proved that the method is reliable for the quantification in each and every matrix (i.e. plasma, kidneys and muscle of chicken) thanks to only one single regression model (i.e. linear) obtained from external calibration standards (without matrix) with deuterated labelled internal standard. The developed method was applied during a depletion study of ceftiofur in chicken tissues and plasma.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
2,2,4-Trimethylpentane, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Heptafluorobutyric acid, ≥99.0% (GC)
Sigma-Aldrich
Heptafluorobutyric acid, 98%
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
SAFC
Iodoacetamide
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
1,4-Dithioerythritol, BioReagent, Molecular Biology, ≥99.0%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets